Order Entry
ContactUsLinkComponent
Pierce™ Amino Acid Modification Reagent, TNBSA, Thermo Scientific
Pierce™ Amino Acid Modification Reagent, TNBSA, Thermo Scientific
Catalog #: CAPI28997
Supplier:  Thermo Scientific
CAS Number:  
TNBSA Solution 100Ml
Pierce™ Amino Acid Modification Reagent, TNBSA, Thermo Scientific
Catalog #: CAPI28997
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    TNBSA solution (2,4,6-trinitrobenzene sulfonic acid), 5% w/v in methanol
  • Size:
    100 mL
  • Cat. no.:
    CAPI28997
  • Supplier No.:
    28997

Specifications

About this item

Thermo Scientific Pierce TNBSA Solution is a 5% solution of trinitrobenzene sulfonic acid in methanol that reacts with primary amines (peptides or amino acids) to yield a soluble colored product, a property useful for various assay methods.

  • TNBSA component reacts readily with primary amino groups of amino acids in aqueous format at pH 8 to form yellow adducts
  • Coloured derivatives are monitored at 345 nm and have extinction coefficients of 10,000 to 15,000
  • No coloured derivatives are formed with secondary amino acids proline and hydroxyproline
  • Applicable to solution or solid phase analysis

TNBSA is used as a hydrophilic modifying reagent for the detection of primary amines in samples containing amino acids, peptides or proteins. It is an excellent reagent for rapid qualitative and quantitative estimation of these biomolecules.

TNBSA is used as a hydrophilic modifying reagent for the detection of primary amines in samples containing amino acids, peptides or proteins. It is an excellent reagent for rapid qualitative and quantitative estimation of these biomolecules.

Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought