Specifications
- Diameter ×L:12 x 75 mm
- Description:Iodination Tube
- Material:Glass
- Size:10 tubes
- Cat. no.:CAPI28601
- Supplier No.:28601
Specifications
About this item
These iodination tubes are pre-coated at the bottom with Pierce™ iodination reagent, which activates iodine-125 for protein or peptide iodination procedures.
- Eliminates the tedious reagent surface coating step
- Consistent, flake-resistant coating of the iodination reagent
- Generates more than sufficient oxidative capacity for a typical protocol
- Allows iodinations to be carried out directly in the tube or in two-steps (see next)
The pre-coated iodination tubes provide for convenient and efficient iodine labeling without the hassles and damaging oxidative effects normally associated with chloramine T and other solution-based methods. Coating glass surfaces to support iodine labeling experiments is difficult to reliably accomplish. These pre-coated glass tubes eliminate that difficult preparative step to ensure that iodination experiments are as simple as possible.