To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Tris-Glycine Buffer, 10X
- Size:1 L
- Cat. no.:CAPI28363
- Supplier No.:28363
Specifications
About this item
These space-saving stock solutions are ideal for quick preparation of standard Tris-glycine (pH 8.5) transfer buffer which is used for Western blotting.
- Easy to use
- Eliminates the possibility of powder remaining in a packet
- Eliminates time spent waiting for powder to dissolve
Required:
Diluted 10-fold in water or 20% methanol, the solution yields 0.025M Tris, 0.192M glycine, pH 8.5
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.