Specifications
- Size:2 x 250 µl
- Storage conditions:Upon receipt store at -20˚C
- Protein/peptide name:Unstained Broad Range Protein Ladder
- Shipping temperature:Product is shipped with an ice pack.
- Cat. no.:CAPI26630
- Supplier No.:26630
Specifications
About this item
Thermo Scientific PageRuler Unstained Broad Range Protein Ladder is a mixture of 11 proteins (5kDa to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.
Features of PageRuler Unstained Broad Range Protein Ladder:
Size range—eleven proteins spanning 5 to 250kDa
Ready-to-use—supplied in a loading buffer for direct loading on gels; no need to boil
Sharp bands—excellent accuracy for detection in with coomassie or silver stains
Reference bands—the 100, 50 and 20kDa bands are more intense for easy orientation
Tagged—each protein in the ladder contains an integral Strep-tag™ II Sequence
Quality tested—each lot evaluated by SDS-PAGE and Western blotting
The protein MW markers in this ladder resolve into clearly identifiable sharp bands ranging in size from 5kDa to 250kDa when analyzed by SDS-PAGE and stained with coomassie dye or silver. The 100, 50 and 20kDa protein bands are of greater intensity and serve as reference bands. Proteins can be detected on Western blots by staining with Ponceau S, coomassie blue or other protein stains. In addition, the ladder proteins (except for the 5kDa peptide) contain an integral Strep-tag™ II Sequence and may be detected on Western blots using Strep-Tactin™ Conjugates. The PageRuler Unstained Broad Range Protein Ladder is supplied ready-to-use: no heating, dilution or addition of a reducing agent is required before use.
Includes:
Proteins in 62.5 mM Tris-H3PO4 (pH 7.5 at 25˚C), 1 mM EDTA, 2% SDS, 100 mM DTT, 1 mM NaN3, 0.01% bromophenol blue and 33% glycerol