Order Entry
Canada
ContactUsLinkComponent
Pierce™ Immunoprecipitation, HA-Tag IP/Co-IP Kit, Thermo Scientific
Pierce™ Immunoprecipitation, HA-Tag IP/Co-IP Kit, Thermo Scientific
Catalog # CAPI26180
Supplier:  Thermo Scientific
CAS Number:  
undefined
Pierce™ Immunoprecipitation, HA-Tag IP/Co-IP Kit, Thermo Scientific
Catalog # CAPI26180
Supplier:  Thermo Scientific
Supplier Number:  26180
CAS Number:  

Specifications

  • Description:
    Pierce™ HA Tag IP/Co-IP kit
  • Size:
    25 reactions
  • Cat. no.:
    CAPI26180
  • Supplier No.:
    26180
  • Sufficient For:
    25 IP/Co-IP Reactions each using 10 µl of Resin

Specifications

About this item

Pierce™ HA-Tag IP/Co-IP kit provides the affinity resin, positive control and other reagents necessary to perform immunoprecipitation (IP) or co-immunoprecipitation (Co-IP) reactions with an HA-tagged bait protein.

  • Highly specific anti-HA monoclonal antibody provides high-yield immunoprecipitation products
  • The kit and anti-HA agarose are compatible with various cell lysates and physiologic buffer systems

This kit is based on crosslinked beaded agarose to which a highly specific anti-HA antibody is covalently immobilised. The ready-to-use affinity resin, together with the included buffers, microcentrifuge spin columns, positive control, and easy-to-follow instructions, constitute a complete set of reagents sufficient for 25 for IP or Co-IP assays.

Delivery information: Kit includes Anti-HA agarose (0.25 ml), HA-tagged positive control (0.5 ml), BupH Tris buffered saline pack (500 ml), Lane Marker Non-reducing Sample Buffer 5X, (5 ml), spin columns accessory pack (27 columns) and collection tubes and caps accessory pack (100 tubes).
Caution: For Research Use Only. Not for use in diagnostic procedures.