Pierce™ Control Agarose Resin, Thermo Scientific
Catalog #: CAPI26150
Supplier: Thermo Scientific
CAS Number:
Specifications
- Description:Control Agarose Resin
- Size:10 mL
- Storage temperature:2 to 8 ˚C
- Cat. no.:CAPI26150
- Supplier No.:26150
Specifications
About this item
Pierce™ Control Agarose Resin is underivatized beaded agarose support for pre-clearing protein immunoprecipitation samples.
- Reduce background signal and non specific binding in IP and Co-IP experiments
- Resin is supplied as a 50% slurry in 20% ethanol (pipetting 1 mL of slurry delivers 0.5 mL of settled resin)
- Crosslinked 4% beaded agarose (CL4B) is the same base support as used in Pierce™ IP and co-IP kits
The resin is effective for depleting proteins and other sample components that bind to agarose nonspecifically, which reduces background signal in IP reactions. When used as a negative control, these beads enable specific and nonspecific binding interactions to be distinguished.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought