Pyridine, Pierce™
Catalog #: CAPI25104
Supplier: Thermo Scientific
CAS Number: 110-86-1
Specifications
- Size:100 g
- Cat. no.:CAPI25104
- Supplier No.:25104
Specifications
Specification Test Results
- Pyridine, Pierce™:
Specification Test Results
About this item
Synonyms: Azabenzene
Formula:C₅H₅N
Boiling Pt:115 °C (1013 hPa)
Melting Pt:–44 °C
Density:0.983 g/cm³ (20 °C)
Flash Pt:17 °C
MDL Number:MFCD00011732
CAS Number:110-86-1
ADR:3,II
:Length
:Width
:Height
:Diameter
:Thickness
:Boiling Pt:
:Boiling Pt: 115 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –44 °C
:Flash Pt:
:Flash Pt: 17 °C
:M.W. 79.1 g/mol
:115 °C (1013 hPa)
UN:1282
:3
:II
:17 °C
:12,08153
MW:79.1 g/mol
:Length:
:Width:
:Height:
:Ø
:Thickness:
:115
:17
Customers Who Bought This Also Bought