ContactUsLinkComponent
Pierce™ Compat-Able™ Protein Assay Preparation Reagent Kit, Thermo Scientific
Pierce™ Compat-Able™ Protein Assay Preparation Reagent Kit, Thermo Scientific
Catalog #: CAPI23215
Supplier:  Thermo Scientific
CAS Number:  
Compat-Able Protein Assay Preparation Reagent Kit
Pierce™ Compat-Able™ Protein Assay Preparation Reagent Kit, Thermo Scientific
Catalog #: CAPI23215
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Compat-Able™ Protein Assay Reagent Kit
  • Size:
    500 mL
  • Cat. no.:
    CAPI23215
  • Supplier No.:
    23215
Specifications
About this item
The Compat-Able™ Protein Assay Preparation Reagent Set eliminates interfering substances from protein assay samples to make them compatible with BCA and Bradford assays.
  • Removes nearly any substance that interferes with colorimetric protein assays: salts, detergents, denaturants, reducing agents, and even IEF sample buffer
  • Prepare samples in less than 10 minutes with four simple steps
  • Reagent set is easier to use than homemade TCA or acetone precipitation reagents, thereby producing more consistent results
  • Microcentrifuge method is easily scaled to larger centrifuge tube volumes for special needs
  • Protocol integrates control and normalization procedures
The Compat-Able Reagents remove salts, detergents, reducing agents and other substances from protein samples to eliminate interference with protein assays. The protocol selectively precipitates all of the protein, thereby allowing the non-protein components to be easily decanted and removed. The purified, precipitated protein is redissolved in water, making it ready for accurate determination of the protein concentration by standard protein assay methods. The procedure is accomplished in less than 10 minutes with just a few simple steps.
Delivery: Kit contains sufficient reagents to pretreat up to 500 samples. It comprises Compat-Able™ Protein Assay Preparation Reagent 1x250 mL and Compat-Able™ Protein Assay Preparation Reagent 2x250 mL.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought