Pierce™ Immobilized p-Aminophenyl Phosphoryl Choline Agarose, Thermo Scientific
Catalog #: CAPI20307
Supplier: Thermo Scientific
CAS Number:
Specifications
- Description:Pierce™ Immobilized p-Aminophenyl Phosphoryl Choline Gel
- Size:5 mL
- Cat. no.:CAPI20307
- Supplier No.:20307
Specifications
About this item
Thermo Scientific Pierce p-Amino Phosphoryl Choline Agarose enables purification of C-reactive protein (CRP) from plasma, ascites and other biological fluids.
- Ligand: p-aminophenyl phosphoryl choline
- Support: crosslinked 6% beaded agarose
- Binding capacity: greater than 3 mg of human C-reactive protein per milliliter of settled resin
- Format: 50% resin slurry in 0.02% sodium azide
- Binding condition: Tris or borate-buffered saline (pH 8 to 8.5) with 1 to 2 mM calcium chloride
- Elution condition: Tris or borate-buffered saline (pH 8 to 8.5) with 2 mM EDTA
- Reusable: resin can be regenerated and reused several times
Caution: For Research Use Only. Not for use in diagnostic procedures. Upon receipt store at 4˚C.
Customers Who Bought This Also Bought