Order Entry
ContactUsLinkComponent
CarboxyLink™ Coupling Resin, Thermo Scientific
CarboxyLink™ Coupling Resin, Thermo Scientific
Catalog #: CAPI20266
Supplier:  Thermo Scientific
CAS Number:  
CarboxyLink Coupling Resin25Ml
CarboxyLink™ Coupling Resin, Thermo Scientific
Catalog #: CAPI20266
Supplier:  Thermo Scientific
CAS Number:  

Specifications

  • Description:
    CarboxyLink™ Coupling Resin
  • Size:
    25 mL
  • Cat. no.:
    CAPI20266
  • Supplier No.:
    20266

Specifications

About this item

Thermo Scientific CarboxyLink Coupling Resin is for covalent immobilization of peptides or other carboxyl-containing (—COOH) molecules to a porous, beaded resin for use in affinity purification procedures.

  • DADPA-activated crosslinked 4% beaded agarose
  • Couple 1 to 2 mg of peptide per mL of resin
  • Immobilizes peptides with high capacity and various orientations without steric hindrance
  • Immoblization results in covalent attachment of carboxyl groups by amide bonds, allowing for multiple rounds of affinity purification with one batch of prepared resin

CarboxyLink Resin is crosslinked beaded agarose that has been activated with diamino-dipropylamine (DADPA) to contain long spacer arms, each with a primary amine at the end. When incubated with the resin and the carbodiimide crosslinker EDC (included in the CarboxyLink Immobilization Kit), carboxyl-containing molecules become permanently attached to the support by stable amide bonds. Once a molecule is coupled, the prepared affinity column can be used multiple times in typical protein affinity purification procedures. CarboxyLink Coupling Resins can also be used to immobilize other kinds of molecules using alternative amine-reactive crosslinking chemistries.

Caution: For Research Use Only. Not for use in diagnostic procedures. Store at 4˚C.
Customers Who Bought This Also Bought