Pierce™ CDI-activated Agarose Resin, Thermo Scientific
Catalog #: CAPI20259
Supplier: Thermo Scientific
CAS Number:
Specifications
- Description:Pierce™ CDI-Activated Agarose Resin
- Size:10 mL
- Cat. no.:CAPI20259
- Supplier No.:20259
Specifications
About this item
Thermo Scientific Pierce CDI-Activated Agarose is carbonyldiimidazole affinity chromatography resin, activated for covalent immobilization of N-nucleophiles and primary amine ligands in aqueous or organic solvent conditions.
- No secondary reagents needed since rection proceeds spontaneously
- Half-life of hydrolysis is much longer than hydroxysuccinimide ester activations
- Reaction is most efficient with primary amines at pH 9 to 11
- Immobilization occurs through the reaction of N-nucleophiles with 1,1' -carbonyldiimidazole groups of the resin to form a stable, uncharged N-alkylcarbamate linkages
- Available in convenient form as it is supplied as stabilized, 50% slurry in acetone
Pierce CDI-activated Resin is porous, beaded agarose chromatography resin that is activated with carbonyl diimidazole groups for covalent immobilization of proteins and other molecules that contain N-nucleophiles. Coupling can be performed in aqueous or organic solvent conditions, making possible the immobilization of many kinds of ligands. Once a molecule is immobilized, the prepared affinity resin can be used multiple times in typical affinity purification procedures.
Caution: For Research Use Only. Not for use in diagnostic procedures. Store at 4˚C.
Customers Who Bought This Also Bought