Pierce™ Iminobiotin Agarose, Thermo Scientific
Catalog #: CAPI20221
Supplier: Thermo Scientific
CAS Number:
Specifications
- Description:Iminobiotin Agarose
- Size:5 mL
- Storage conditions:4 ˚C
- Cat. no.:CAPI20221
- Supplier No.:20221
Specifications
About this item
Thermo Scientific Pierce Iminobiotin Agarose provides for unusual affinity purification experiments involving clean-up or removal of streptavidin or avidin protein components.
- Crosslinked beaded agarose onto which iminobiotin is covalently immobilized
- Use Iminobiotin Agarose for reversible binding (e.g., non-denaturing purification of avidin)
Immobilized iminobiotin is the guanido analog of biotin. The dissociation constant of the avidin-iminobiotin complex is pH dependent. At pH 9.5 to 11, the avidin-iminobiotin complex will bind tightly. At pH 4, the avidin-iminobiotin complex will dissociate. Because no denaturing agents such as 8 M guanidine hydrochloride or 4 M urea are required for elution, the avidin conjugate has a better chance of maintaining its activity after purification. Thus, iminobiotin agarose can be used for situations requiring reversible binding and elution of avidin components.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought