Pierce™ Immobilized GST Agarose, Thermo Scientific
Catalog #: CAPI20211
Supplier: Thermo Scientific
CAS Number:
Specifications
- Description:Immobilized GST Agarose
- Size:5 mL
- Storage conditions:4 ˚C
- Cat. no.:CAPI20211
- Supplier No.:20211
Specifications
About this item
Thermo Scientific Immobilized GST is glutathione S-transferase from Schistosoma japonicum that has been covalently immobilized on crosslinked, 6% beaded agarose.
- Provides purification of specific antibodies from those directed against the GST tag
- Bound antibodies can be eluted with 6 M guanidinium chloride allowing for 10 purifications with the support
Antibody molecules recognizing the GST portion of a recombinant fusion protein can be removed from the desired antibody by passing antiserum over an immobilized GST column
The expression of foreign proteins in E. coli using a glutathione S-transferase (GST) fusion system is a method of choice for studying the function of particular proteins, for purifying interacting factors and for generating antibodies against fused proteins.
GST antibodies will bind to the immobilized GST column, whereas antibodies against other fusion proteins will pass through the column unrestrained.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought