Specifications
- Length:11"
- Description:Uterine Curette Set
- Cat. no.:10194-002
- Supplier No.:90-5428
Specifications
About this item
Sims uterine curette set is a collection of 6 OB/GYN curettes used for removing tissue from the lining of the uterus.
- Stainless steel
They may be used to scrape and remove growths or tissue to be biopsied. They may also be used as part of a D and C procedure. The teardrop shaped, fenestrated, loop tip is sharp and available in six sizes in this set. The shaft is malleable so that it can be manipulated to an angle suitable for both user and patient needs.