Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Supplier: VWR International
Certificates
About this item
Synonyms: 2-Propanone,DMK,Propanone
Formula:CH₃COCH₃
MW:58.08 g/mol
Boiling Pt:56.2 °C (1013 hPa)
Melting Pt:–95.4 °C
Density:0.792 g/cm³ (20 °C)
MDL Number:MFCD00008765
CAS Number:67-64-1
Merck Index:13,00067
:Storage Temperature: Ambient
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
Flash Pt:<–20 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 56.2 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –95.4 °C
:Flash Pt:
:Flash Pt: <–20 °C (closed cup)
:M.W. 58.08 g/mol
EINECS:200-662-2
:56.2
UN:1090
REACH:01-2119471330-49
:3
:II
:<–20 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (by GC-FID) (Corrected for Water):Min. 99.5 %
- Acidity:Max. 0.0005 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Residue after evaporation:Max. 5 ppm
- Water (by KF coulometry):Max. 0.5 %
- Transmittance (345 nm):Min. 50 %
- Transmittance (350 nm):Min. 80 %
- Transmittance (360 nm):Min. 98 %
- VWR Chemicals BDH®, Acetone ≥99.5%, HiPerSolv CHROMANORM® for HPLC:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals VWR Chemicals BDH® HiPerSolv CHROMANORM® Solvents for HPLC & LC-MS
Product Literature
BDH20067.400 Acetone Product specification
Product specification
Customers Who Bought This Also Bought