Gracey Curette, OR Grade, Sklar
Supplier: Sklar
Certificates
About this item
Gracey periodontal curette is a double-ended dental curette, used to scale and remove deposits from posterior teeth.
The blade variations are specific to the particular posterior tooth surface being cleaned.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.