Formvar Carbon Support Film on Square Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
The formvar coated grids are stabilized with an evaporated carbon film.
- Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
- Carbon coatings thickness of 1 nm, 3 to 4 nm, and 20 to 30 nm
- Select copper, nickel, or gold grids in 100, 150, 200, 300, or 400 mesh
- Optically checked followed by batch testing in the EM
This type of coating is excellent for specimen support, especially for ultra thin sections. All film is laid on the shiny side of the grid.
Packaging: Packed in grid storage box.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.