Sodium chloride 99.5-100.5% (by argentometric analysis, on dry substance), EMPROVE® EXPERT Ph. Eur., BP, JP, USP suitable for use as excipient low in endotoxins, SAFC®
Supplier: EMD MILLIPORE – PCS CA EXCEP
Certificates
About this item
Synonyms: Saline,Sea salt
Formula:NaCl
MW:58.44 g/mol
Boiling Pt:1461 °C (1013 hPa)
Melting Pt:801 °C
Density:2.17 g/cm³ (20 °C)
MDL Number:MFCD00003477
CAS Number:7647-14-5
:Sodium chloride
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5-100,5
:Boiling Pt:
:Boiling Pt: 1461 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 801 °C
:Flash Pt:
:Flash Pt:
:M.W. 58.44 g/mol
:1461 °C (1013 hPa)
Storage Temperature:Ambient
:12,08742
:14,08599
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (argentometric; calculated on dried substance):99.5 - 100.5 %
- Identity:passes test
- Appearance of solution:passes test
- Acidity or alkalinity:passes test
- Bromide (Br):≤ 0.005 %
- Ferrocyanides:passes test
- Iodide (I):passes test
- Nitrite (NO₂) (UV-absorption 354nm; 1cm; 10%; water):≤ 0.01
- Phosphate (PO₄):≤ 0.0025 %
- Sulfate (SO₄):≤ 0.0200 %
- Heavy metals (as Pb):≤ 0.0003 %
- Al (Aluminium):≤ 0.00002 %
- As (Arsenic):≤ 0.0001 %
- Ba (Barium):passes test
- Ca (Calcium):≤ 0.002 %
- Fe (Iron):≤ 0.0002 %
- K (Potassium):≤ 0.01 %
- Magnesium and alkaline-earth metals (as Ca):≤ 0.0100 %
- Residual solvents (ICH Q3C):excluded by production process
- Loss on drying (105 °C, 2 h):≤ 0.5 %
- Endotoxins (kinetic turbidimetric test):≤ 2.5 I.U./g
- SAFC®, Sodium chloride 99.5-100.5% (by argentometric analysis, on dry substance), EMPROVE® EXPERT Ph. Eur., BP, JP, USP suitable for use as excipient low in endotoxins:
Specification Test Results
Customers Who Bought This Also Bought