Sodium Cacodylate Buffer, Electron Microscopy Sciences
Supplier: Electron Microscopy Science
Certificates
About this item
Used for preparing buffered glutaraldehdye fixatives for electron microscopy
Available with a pH of 7.2 or 7.4.
Sodium Cacodylate Buffer, 0.2M is prepared from Sodium Cacodylate Trihydrate (F.W 214.02).
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.