Specifications
- Description:Sterile cell phone cover
- Material:Ultra-clear polyethylene
- Cat. no.:10798-962
- Supplier No.:EC2100
Specifications
About this item
eShield™ sterile cell phone covers are designed to protect a sterile environment from contaminants on smart phones.
- Covers are touchscreen compatible, even with double gloves
- Phone covers easily accommodate aseptic technique
They are constructed of ultra-clear polyethylene film ensuring clear photos.
The sterile barrier provided by eShield™ covers enables the medical professional to use a smart phone to access studies, record data, conduct consultations, and capture photos conveniently and safely without contaminating the sterile environment.
They can be used within the sterile field in any medical environment, including surgical suites and clinical and research labs.