To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Actin Beta
- Clonality:Polyclonal
- Gene ID:ACTB/280979/442937/396526/100135470/60/443052/414396/100009272; Actb/11461/81822
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse,Cat,Drosophila,Pig,Rabbit,Leech,Sheep,Chicken,Human,Bovine,Guinea pig,Dog,Rat
- Antigen symbol:ACTB
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:500 µL
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:PS1TP5-binding protein 1|cytoplasmic 1|PS1TP5BP1|BRWS1|beta cytoskeletal actin|actin
- Storage buffer:PBS, pH 7.6, with 0.2% BSA and 15mM Sodium azide
- Storage temperature:4 °C
- Tested applications:Immunohistochemistry (Paraffin)
- Cat. no.:CAPIPA5-16914
- Supplier No.:PA5-16914
Specifications
About this item
PA5-16914 targets Actin Beta in IHC (P) and WB applications and shows reactivity with Bovine, Canine, Chicken, Drosophila, Feline, Guinea Pig, Human, Leech, mouse, Ovine, Porcine, Rabbit, and Rat samples. The PA5-16914 immunogen is synthetic peptide corresponding to N-terminus of human beta-actin. The two major cytoskeletal proteins implicated in cell motility are actin and myosin. There are three alpha-actin (skeletal, cardiac, and smooth muscle), one beta-actin (beta nonmuscle), and two gamma-actins (gamma-smooth nuscle and gamma-nonmuscle). Antibody to beta-actin provides a specific and useful tool in studying the intracellular distribution of beta-actin and the static and dynamic aspects of the cytoskeleton.
- Immunohistochemistry (Paraffin): 1:50-1:100, Western Blot: 2-4 µg/ml
Type: Primary
Antigen: beta Actin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Guinea Pig, Canine, Human, Drosophila, Chicken, Bovine, Feline, Rabbit, Leech, Ovine, Porcine, Mouse, Rat