To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Alcohol Dehydrogenase 5
- Clonality:Polyclonal
- Gene ID:ADH5/128; Adh5/11532/100145871
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:ADH5
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:glutathione-dependent formaldehyde dehydrogenase|class III alcohol dehydrogenase 5 chi subunit|GSNOR|Alcohol dehydrogenase (class III)|formaldehyde dehydrogenase|alcohol dehydrogenase 5 (class III)|ADH-3|ADH5|FDH |chi polypeptide|ADHX
- Storage buffer:TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Tested applications:Immunohistochemistry
- Cat. no.:CAPIPA5-18651
- Supplier No.:PA5-18651
Specifications
About this item
This antibody is predicted to react with bovine, canine and porcine based on sequence homology. This gene encodes a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The encoded protein forms a homodimer. It has virtually no activity for ethanol oxidation, but exhibits high activity for oxidation of long-chain primary alcohols and for oxidation of S-hydroxymethyl-glutathione, a spontaneous adduct between formaldehyde and glutathione. This enzyme is an important component of cellular metabolism for the elimination of formaldehyde, a potent irritant and sensitizing agent that causes lacrymation, rhinitis, pharyngitis, and contact dermatitis. The human genome contains several non-transcribed pseudogenes related to this gene.
Immunohistochemistry: Assay Dependent, Western Blot: 0.5-1.5 ug/ml
Type: Primary
Antigen: ADH5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Human, Rat