To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Gap Junction Protein, Beta 2
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:GJB2
- Conjugation:Unconjugated
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:CX26|DFNB1A|DFNB1|NSRD1|HID|DFNA3A|PPK|DFNA3|KID|connexin 26|Gap junction beta-2 protein
- Storage buffer:TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Cat. no.:CAPIPA5-18618
- Supplier No.:PA5-18618
Specifications
About this item
This antibody is predicted to react with human and rat based on sequence homology. This gene encodes a member of the gap junction protein family. The gap junctions were first characterized by electron microscopy as regionally specialized structures on plasma membranes of contacting adherent cells. These structures were shown to consist of cell-to-cell channels that facilitate the transfer of ions and small molecules between cells. The gap junction proteins, also known as connexins, purified from fractions of enriched gap junctions from different tissues differ. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene are responsible for as much as 50% of pre-lingual, recessive deafness.
Western Blot: 0.01-0.03 ug/ml
Type: Primary
Antigen: CX26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse