To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Peptidylprolyl Cis/trans Isomerase NIMA-interacting 1
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Rat
- Antigen symbol:PIN1
- Conjugation:Unconjugated
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:Peptidyl-prolyl cis-trans isomerase NIMA-interacting 1|Rotamase Pin1|EC 5.2.1.8|UBL5|DOD|PPIase Pin1
- Storage buffer:TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Cat. no.:CAPIPA5-18594
- Supplier No.:PA5-18594
Specifications
About this item
This antibody is predicted to react with canine, human and mouse based on sequence homology. Peptidyl-prolyl cis/trans isomerases , such as PIN1, catalyze the cis/trans isomerization of peptidyl-prolyl peptide bonds. PIN1 is the only PPIase that specifically binds to phosphorylated ser/thr-pro motifs to catalytically regulate the post-phosphorylation conformation of its substrates. PIN1-catalyzed conformational regulation has a profound impact on key proteins involved in the regulation of cell growth, genotoxic and other stress responses, the immune response, germ cell development, neuronal differentiation, and survival .
Recommended Dilutions: Western Blot: 0.2-0.6 ug/ml
Type: Primary
Antigen: PIN1 (peptidylprolyl cis/trans isomerase, NIMA-interacting 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rat