To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 9
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:ZNF9
- Conjugation:Unconjugated
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:CNBP1|CNBP|zinc finger protein 9|DM2|zinc finger protein 9 (a cellular retroviral nucleic acid binding protein) |Proximal myotonic myopathy nucleic acid binding protein|RNF163 |PROMM|HGNC:13164|zinc finger protein 273|ZNF9
- Storage buffer:TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Cat. no.:CAPIPA5-18505
- Supplier No.:PA5-18505
Specifications
About this item
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This gene encodes a nucleic-acid binding protein with seven zinc-finger domains. The protein has a preference for binding single stranded DNA and RNA. The protein functions in cap-independent translation of ornithine decarboxylase mRNA, and may also function in sterol-mediated transcriptional regulation. A CCTG expansion in the first intron of this gene results in myotonic dystrophy type 2. Multiple transcript variants encoding different isoforms have been found for this gene.
Recommended Dilutions: Western Blot: 1-3 ug/ml
Type: Primary
Antigen: CNBP (CCHC-type zinc finger, nucleic acid binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human