To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:HEXIM1
- Conjugation:Unconjugated
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:hexamethylene bisacetamide-inducible protein|hexamthylene bis-acetamide inducible 1|CLP1|ED|hexamethylene-bis-acetamide-inducible transcript 1|hexamethylene bis-acetamide inducible 1|menage a quatre 1|HMBA-inducible|MAQ1 |HEXIM1|HIS1|HGNC:24953
- Storage buffer:TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Cat. no.:CAPIPA5-18434
- Supplier No.:PA5-18434
Specifications
About this item
This antibody is predicted to react with mouse based on sequence homology. Expression of this gene is induced by hexamethylene-bis-acetamide in vascular smooth muscle cells. This gene has no introns.
Recommended Dilutions: Western Blot: 0.3-1 ug/ml
Type: Primary
Antigen: HEXIM1 (hexamethylene bis-acetamide inducible 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human