To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Low Density Lipoprotein Receptor-related Protein 5
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:LRP5
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Cross adsorption:No
- Antigen synonyms:LRP7|EVR4|OPS|OPPG|BMND1|HBM|LR3|VBCH2|EVR1|OPTA1|LRP-5
- Storage buffer:TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Tested applications:Immunohistochemistry
- Cat. no.:CAPIPA5-18387
- Supplier No.:PA5-18387
Specifications
About this item
This antibody is predicted to react with mouse and rat based on sequence homology. This gene encodes a transmembrane low-density lipoprotein receptor that binds and internalizes ligands in the process of receptor-mediated endocytosis. This protein also acts as a co-receptor with Frizzled protein family members for transducing signals by Wnt proteins and was originally cloned on the basis of its association with type 1 diabetes mellitus in humans. This protein plays a key role in skeletal homeostasis and many bone density related diseases are caused by mutations in this gene. Mutations in this gene also cause familial exudative vitreoretinopathy.
Recommended Dilutions: Immunohistochemistry: 3-6µg/ml
Type: Primary
Antigen: LRP5 (low density lipoprotein receptor-related protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human