To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD2 (cytoplasmic tail) binding protein 2
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CD2BP2
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:CD2 binding protein 2|U5-52K|FWP010|CD2 cytoplasmic domain binding protein 2|U5 snRNP 52K protein|KIAA1178|CD2 antigen cytoplasmic tail-binding protein 2|Snu40|LIN1|CD2 cytoplasmic tail binding protein 2|CD2 antigen cytoplasmic tail binding protein 2|CD2BP2
- Storage buffer:TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Tested applications:Immunohistochemistry (Paraffin)
- Cat. no.:CAPIPA5-18286
- Supplier No.:PA5-18286
Specifications
About this item
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This gene encodes a bi-functional protein. In the cytoplasm, the encoded protein binds the cytoplasmic tail of human surface antigen CD2 via its C-terminal GYF domain, and regulate CD2-triggered T lymphocyte activation. In the nucleus, this protein is a component of the U5 small nuclear ribonucleoprotein complex and is involved in RNA splicing. A pseudogene has been identified on chromosome 7. Alternative splicing results in multiple transcript variants but their biological validity has not been determined.
Immunohistochemistry (Paraffin): 5-10 ug/ml, Western Blot: 0.3-1.0 ug/ml
Type: Primary
Antigen: CD2BP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human