To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Melanoma Differentiation-Associated protein 5
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:MDA5
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:Clinically amyopathic dermatomyositis autoantigen 140 kDa|RLR-2|Helicard|Murabutide down-regulated protein|Melanoma differentiation-associated protein 5|RNA helicase-DEAD box protein 116|MDA-5|CADM-140 autoantigen|RIG-I-like receptor 2|Helicase with 2 CARD domains|Interferon-induced with helicase C domain protein 1
- Storage buffer:TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Tested applications:Immunohistochemistry
- Cat. no.:CAPIPA5-18164
- Supplier No.:PA5-18164
Specifications
About this item
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp and a protein kinase C-activating compound, mezerein . Irreversible reprogramming of melanomas can be achieved by treatment with both these agents; treatment with either agent alone only achieves reversible differentiation.
Recommended Dilutions: Western Blot: Assay-dependent, Immunohistochemistry: 10µg/ml
Type: Primary
Antigen: IFIH1 (interferon induced with helicase C domain 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human