To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Capping protein (actin filament) muscle Z-line, beta
- Clonality:Polyclonal
- Gene ID:CAPZB/832/100037959; Capzb/12345/298584
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Human,Rat,Mouse
- Antigen symbol:CAPZB
- Conjugation:Unconjugated
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:CAPPB|F-actin capping protein beta subunit|Cap Z|CAPB|RP4-657E11.7|MGC104401|MGC129749|MGC129750|CAPZ
- Storage buffer:TBS, pH 7.3, with 0.5% BSA and 0.02% Sodium azide
- Storage temperature:–20 °C, avoid freeze/thaw cycles
- Cat. no.:CAPIPA5-17961
- Supplier No.:PA5-17961
Specifications
About this item
This antibody is predicted to react with bovine and canine based on sequence homology. CAPZB is a member of the F-actin capping protein family. This gene encodes the beta subunit of the barbed-end actin binding protein. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
Western Blot: 0.1-0.3 µg/ml
Type: Primary
Antigen: CAPZB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Porcine, Human, Rat