To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:AKT Interacting Protein
- Clonality:Polyclonal
- Host:Mouse
- Reactivity:Human
- Antigen symbol:AKTIP
- Conjugation:Unconjugated
- Size:50 µg
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:FT1|FTS
- Storage buffer:In 1x PBS, pH 7.4
- Sequence:MNPFWSMSTSSVRKRSEGEEKTLTGDVKTSPPRTAPKKQLPSIPKNALPITKPTSPAPAAQSTNGTHASYGPFYLEYSLLAEFTLVVKQKLPGVYVQPSYRSALMWFGVIFIRHGLYQDGVFKFTVYIPDNYPDGDCPRLVFDIPVFHPLVDPTSGELDVKRAFAKWRRNHNHIWQVLMYARRVFYKIDTASPLNPEAAVLYEKDIQLFKSKVVDSVKVCTARLFDQPKIEDPYAISFSPWNPSVHDEAREKMLTQKKPEEQHNKSVHVAGLSWVKPGSVQPFSKEEKTVAT
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:AKTIP (NP_071921.1, 1 a.a. ~ 292 a.a) full-length human protein
- Cat. no.:10509-586
- Supplier No.:H00064400-B01P
Specifications
About this item
Mouse polyclonal antibody raised against a full-length human AKTIP protein.
Recommended Dilutons: Western Blot (1 ug/mL)
Type: Primary
Antigen: AKTIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human