To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- ChIP:Yes
- Clonality:Polyclonal
- Gel shift:Yes
- Gene ID:FGFR2/2263; Fgfr2/14183/25022
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Primates,Human,Rat,Mouse
- Antigen symbol:FGFR2/CD332
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:TK14|FGF Receptor 2|ECT1|TK25|BEK|JWS|KGFR|CFD1|CEK3|CD332|Fibroblast Growth Factor Receptor 2|K-SAM|BFR-1
- Storage buffer:PBS with 0.09% Sodium azide
- Molecular weight:92.03 kDa
- Storage temperature:Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at –20 °C
- Concentration:Lot-specific
- Immunogen:KLH conjugated synthetic peptide between 7-37 amino acids from the N-terminal region of human FGFR2
- Tested applications:Immunohistochemistry
- Purification:Protein G, Size-exclusion - Dialysis
- Cat. no.:CAPIPA5-14651
- Supplier No.:PA514651
Specifications
About this item
FGFR2 is a member of the fibroblast growth factor receptor family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein consists of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. This particular family member is a high-affinity receptor for acidic, basic and/or keratinocyte growth factor, depending on the isoform. Mutations in the gene are associated with many craniosynostotic syndromes and bone malformations. The genomic organization of the gene encompasses 20 exons. Alternative splicing in multiple exons, including those encoding the Ig-like domains, the transmembrane region and the carboxyl terminus, results in varied isoforms which differ in structure and specificity. Isoform 1 has equal affinity for aFGF and bFGF but does not bind KGF.
Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:50-1:100, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: FGFR2 (fibroblast growth factor receptor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: