To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ephrin type A receptor 2
- ChIP:Yes
- Clonality:Polyclonal
- Gel shift:Yes
- Gene ID:EPHA2/1969; Epha2/13836
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human,Mouse
- Antigen symbol:EPHA2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Epithelial cell kinase|CTPA|ARCC2|ECK|CTRCT6|CTPP1|Ephrin type-A receptor 2|Tyrosine-protein kinase receptor ECK
- Storage buffer:PBS with 0.09% Sodium azide
- Molecular weight:108.25 kDa
- Storage temperature:Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at –20 °C
- Concentration:Lot-specific
- Immunogen:KLH conjugated synthetic peptide between 30-60 amino acids from the N-terminal region of human EphA2
- Tested applications:Immunohistochemistry
- Purification:Ammonium sulfate precipitation, Size-exclusion - Dialysis
- Cat. no.:CAPIPA5-14574
- Supplier No.:PA514574
Specifications
About this item
This antibody is predicted to react with non-human primate based on sequence homology. Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: EPHA2 (EPH receptor A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: