To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:FK506-Binding Protein 10
- ChIP:Yes
- Clonality:Polyclonal
- Gel shift:Yes
- Gene ID:FKBP10/60681; Fkbp10/14230
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human,Mouse
- Antigen symbol:FKBP10
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:Immunophilin FKBP65|FKBP65|65 kDa FK506-binding protein|FKBP10|PSEC0056|Rotamase|Peptidyl-prolyl cis-trans isomerase|FK506-binding protein 10|PPIase
- Storage buffer:PBS with 0.09% Sodium azide
- Molecular weight:64.24 kDa
- Storage temperature:Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at –20 °C
- Concentration:Lot-specific
- Immunogen:KLH conjugated synthetic peptide between 57-87 amino acids from the N-terminal region of human FKBP10
- Tested applications:Immunohistochemistry
- Purification:Antigen affinity chromatography
- Cat. no.:CAPIPA5-14272
- Supplier No.:PA514272
Specifications
About this item
The protein belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones.
Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: FKBP10 (FK506 binding protein 10, 65 kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human