To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase D1
- ChIP:Yes
- Clonality:Polyclonal
- Gel shift:Yes
- Gene ID:PRKD1/5587; Prkd1/18760
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human,Mouse
- Antigen symbol:PRKD1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:PRKCM|Protein kinase D|nPKC-D1|PKD|Serine/threonine-protein kinase D1|PKD1|Protein kinase C mu type|PRKD1|nPKC-mu
- Storage buffer:PBS with 0.09% Sodium azide
- Molecular weight:101.89 kDa
- Storage temperature:Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at –20 °C
- Concentration:Lot-specific
- Immunogen:KLH conjugated synthetic peptide between 361-389 amino acids from the N-terminal region of human PKC mu
- Tested applications:Immunohistochemistry
- Purification:Ammonium sulfate precipitation, Size-exclusion - Dialysis
- Cat. no.:CAPIPA5-13749
- Supplier No.:PA513749
Specifications
About this item
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play distinct roles in cells. PKC mu is one of the PKC family members. The protein is a cytosolic serine-threonine kinase that binds to the trans-Golgi network and regulates the fission of transport carriers specifically destined to the cell surface.
Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100
Type: Primary
Antigen: PRKD1 (protein kinase D1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: