To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Glutamate Receptor, Metabotropic 5
- ChIP:Yes
- Clonality:Polyclonal
- Gel shift:Yes
- Gene ID:GRM5/2915; Grm5/108071
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human,Mouse
- Antigen symbol:GRM5
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:GPRC1E|MGLUR5|GRM5|Metabotropic glutamate receptor 5 [Precursor]|mGluR5
- Storage buffer:PBS with 0.09% Sodium azide
- Molecular weight:128.87 kDa
- Storage temperature:Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at –20 °C
- Concentration:Lot-specific
- Immunogen:KLH conjugated synthetic peptide between 1020-1050 amino acids from the C-terminal region of human Metabotropic Glutamate Receptor 5 (GPRC1E)
- Purification:Ammonium sulfate precipitation, Size-exclusion - Dialysis
- Cat. no.:CAPIPA5-13361
- Supplier No.:PA513361
Specifications
About this item
L-glutamate is the major excitatory neurotransmitter in the central nervous system and activates both ionotropic and metabotropic glutamate receptors. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein-coupled receptors, that have been divided into 3 groups on the basis of sequence homology, putative signal transduction mechanisms, and pharmacologic properties. Group I includes GRM1 and GRM5 (also known as GPRC1E) and these receptors have been shown to activate phospholipase C. Group II includes GRM2 and GRM3 while Group III includes GRM4, GRM6, GRM7 and GRM8. Group II and III receptors are linked to the inhibition of the cyclic AMP cascade but differ in their agonist selectivities. The activity of GRM5 is mediated by a G-protein that activates a phosphatidylinositol-calcium second messenger system and generates a calcium-activated chloride current.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: GRM5 (glutamate receptor, metabotropic 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: