To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Septin 9
- ChIP:Yes
- Clonality:Polyclonal
- Gel shift:Yes
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:SEPT9
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:PNUTL4|SeptD1|MSF|SINT1|NAPB|MSF1|AF17q25
- Storage buffer:PBS with 0.09% Sodium azide
- Molecular weight:63.63 kDa
- Storage temperature:Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at –20 °C
- Concentration:Lot-specific
- Immunogen:KLH conjugated synthetic peptide between 57-85 amino acids from the C-terminal region of human SEPT9
- Tested applications:Immunohistochemistry
- Purification:Ammonium sulfate precipitation, Size-exclusion - Dialysis
- Cat. no.:CAPIPA5-13200
- Supplier No.:PA513200
Specifications
About this item
The maf oncogene was identified by structural analysis of the AS42 avian transforming retrovirus genome. The Maf family is divided into two subclasses, large Mafs (vMaf, cMaf, MafB and Nrl) and small Mafs (MafF, MafK, and MafG). Both subclasses contain leucinezipper motifs, which allow homodimerization as well as heterodimerization with a variety of other bZip transcription factors. Large Mafs also contain an acidic transactivation domain absent in the small Maf proteins. Although they do not possess inherent transactivation activity, small Maf proteins can act as positive regulators of transcription by targeting transcriptionally active dimerization partners to specific DNA regulatory elements. Conversely, small Mafs can act also as negative regulators of transcription by recruiting transcriptional repressors or by forming homodimers that can replace active dimers. Human MafF was isolated in a yeast one-hybrid system from a human myometrium cDNA library. Human MAFF encodes a 164 amino acids proten. Like other small MAFF proteins, it contains an extended leucine zipper structure and lacks an N-terminal transactivating domain. The three small Maf proteins have been implicated in a number of physiological processes, including development, differentiation, haematopoiesis and stress response. Interestingly, these three proteins regulate the stress response via different mechanisms.
Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50
Type: Primary
Antigen: MAFK (v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog K)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human