To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:RAD9 Checkpoint Clamp Component A
- ChIP:Yes
- Clonality:Polyclonal
- Gel shift:Yes
- Host:Rabbit
- Isotype:IgG
- Neutralisation:Yes
- Reactivity:Human
- Antigen symbol:RAD9
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Size:400 µL
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:RAD9 homolog|RAD 9 (S pombe) homolog|RAD9A|Cell cycle checkpoint control protein|RAD9 homolog A
- Modification:phospho-Tyr28
- Storage buffer:PBS with 0.09% Sodium azide
- Molecular weight:42.55 kDa
- Storage temperature:Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at –20 °C
- Concentration:Lot-specific
- Immunogen:KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding Y28 of human RAD9
- Purification:Antigen affinity chromatography
- Cat. no.:CAPIPA5-12676
- Supplier No.:PA512676
Specifications
About this item
This antibody is predicted to react with non-human primate based on sequence homology. Rad9 is highly similar to Schizosaccharomyces pombe rad9, a cell cycle checkpoint protein required for cell cycle arrest and DNA damage repair in response to DNA damage. This protein is found to possess 3' to 5' exonuclease activity, which may contribute to its role in sensing and repairing DNA damage. It forms a checkpoint protein complex with RAD1 and HUS1. This complex is recruited by checkpoint protein RAD17 to the sites of DNA damage, which is thought to be important for triggering the checkpoint-signaling cascade.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: RAD9A (RAD9 homolog A (S. pombe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human