Specifications
- CAS No.:128270-60-0
- Size:100 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD08692016
- Protein/peptide name:Bivalirudin Trifluoroacetate
- Molecular weight:2180.32
- Sequence:H-D-Phe-Pro-Arg-Pro-Gly-Gly-Gly-Gly-Asn-Gly-Asp-Phe-Glu-Glu-Ile-Pro-Glu-Glu-Tyr-Leu-OH
- Molecular formula:C₉₈H₁₃₈N₂₄O₃₃
- Cat. no.:H-6668.0100BA
- Supplier No.:H-6668.0100
Specifications
About this item
Bivalirudin is a highly specific and reversible direct thrombin inhibitor. Offered under Bolar Exemption: All APIs that are sold for development of drug products still patent protected are offered under Bolar Exemption only. The following disclaimer applies: These products are offered and sold in small quantities only and solely for uses reasonably related to privileged trials and studies for obtaining marketing authorization required by law (Bolar Exemption). Bachem cannot be made liable for any infringement of intellectual property rights. It is the sole and only responsibility of the purchaser or user of these products to comply with the relevant national rules and regulations.
These peptides correspond to the C-terminus of the 65-residue polypeptide hirudin, a potent thrombin inhibitor found in the saliva of the leech Hirudo medicinalis. Treatment of hirudin with carboxypeptidase Y has shown that the last 5 amino acids are essential for its inhibitory activity.