Human Acetyl-Tetracosactide (4-24) Trifluoroacetate
Catalog #: H-6748.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Species:Human
- Size:1 mg
- Storage conditions:–20 ± 5 °C
- MDL No.:MFCD18782471
- Protein/peptide name:Acetyl-Tetracosactide (4-24) Trifluoroacetate
- Molecular weight:2638.19
- Sequence:Ac-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-Gly-Lys-Lys-Arg-Arg-Pro-Val-Lys-Val-Tyr-Pro-OH
- Molecular formula:C₁₂₃H₁₉₃N₃₇O₂₆S
- Cat. no.:H-6748.0001BA
- Supplier No.:H-6748.0001
Specifications
About this item
Please see also α-MSH (corresponds to acetyl-ACTH (1-13) amide, H-1075), α-MSH (free acid) (acetyl-ACTH (1-13), H-1070), and (Des-acetyl)-α-MSH (H-4390). Wied: Pituitary Adrenal System Hormones and Behaviour. Symposium on Developments in Endocrinology (1976) / Anon.: ACTH and Related Peptides: Structure, Regulation, and Action. Ann. N.Y. Acad. Sci. 297, 1 (1977) / A.V.Schally: Aspects of Hypothalamic Regulation of the Pituitary Gland. Science 202, 18 (1978) / R.Schwyzer: Studies on Polypeptide Receptors. A Critical View on the Mechanism of ACTH Action. Bull. Schweiz. Acad. Med. Wiss. 34, 263 (1978).
Customers Who Bought This Also Bought