To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:SWI/SNF Related Matrix Associated Actin Dependent Regulator of Chromatin Subfamily D Member 3
- Clonality:Polyclonal
- Gene ID:6604
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SMARCD3
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CRACD3|BAF60C|Rsc6p
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10490-218
- Supplier No.:BS-9479R
Specifications
About this item
SMARCD3, is a member of the SMARCD family and contains one SWIB domain. Two isoforms, isoform 1 and isoform 2 exist due to alternative splicing events. Both isoforms are expressed in placenta, salivary gland, kidney, brain, trachea, uterus, prostate, testis, thyroid, spleen and heart, while isoform 1 is also expressed in adipose tissue and skeletal muscle. Localizing to the nucleus, SMARCD3 is a component of the ATP-dependent chromatin remodeling complex SNF/SWI and is believed to play a role in nucleosome remodeling. SMARCD3 also plays an important role in the regulation of muscle development. In mice, the silencing of the gene en-coding SMARCD3 leads to defects in heart morphogenesis. In addition, both isoforms of SMARCD3 directly interact with and function as coactivators for several transcription factors.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: SMARCD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat