To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 19S 10B
- Clonality:Polyclonal
- Gene ID:5706
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:Proteasome 19S 10B
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:26S protease regulatory subunit 10B|PSMC6|Proteasome 26S subunit ATPase 6|SUG2.|26S proteasome AAA-ATPase subunit RPT4|PRS10_HUMAN|Proteasome subunit p42|PRS10
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10489-166
- Supplier No.:BS-9346R
Specifications
About this item
The 26S proteasome is a large complex involved in the intracellular degradation of proteins in eukaryotes. Ubiquitination by E3 ubiquitin ligases targets proteins for degradation by this complex. The 26S proteasome plays an important role in the regulation of many biological processes. It is composed of over 30 different subunits and contains at least two copies of each subunit. Assembly of this large complex is ATP-dependent. Due to its size, it is fairly unstable and often disassociates into subcomplexes (including a 20S core and two 19S regulatory complexes). The 26s proteasome p42A (also known as Rpn7 in yeast and S10 in human) is one of at least nine, non-ATPase lid subunits of the 19S regulatory complex. It is important in the proper assembly and stability of the 26S proteasome. The 19S regulatory complex recognizes ubiquitinated proteins, removes the ubiquitin chains and translocates the proteins to the 20S core for degradation.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: Proteasome 19S 10B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat