To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Ring Finger Protein 23
- Clonality:Polyclonal
- Gene ID:56658
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:RNF23
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:TFP RING FINGER PROTEIN 23|TFP|Ring finger protein 23|Tripartite mot containing 39|RNF23|Testis abundant finger protein|TRI39_HUMAN.|TRIM39
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10669-724
- Supplier No.:BS-9238R
Specifications
About this item
The RING-type zinc finger motif is present in a number of viral and eukaryotic proteins and is made of a conserved cysteine-rich domain that is able to bind two zinc atoms. Proteins that contain this conserved domain are generally involved in the ubiquitination pathway of protein degradation. RNF23 (RING finger protein 23), also known as tripartite motif-containing protein 39 (TRIM39) or testis-abundant finger protein, is a 518 amino acid protein belonging to the TRIM/RBCC family that is known to interact with MOAP1. Ubiquitously expressed and existing as two alternatively spliced isoforms, RNF23 is found at highest levels in spleen, testis, brain, kidney, liver, heart and skeletal muscle. RNF23 typically localizes to cytosol but shifts to mitochondria upon co-localization with MOAP1, a short-lived, pro-apoptotic protein which RNF23 prevents from becoming poly-ubiquitinated and degraded, thereby facilitating apoptosis. RNF23 contains one B box-type zinc finger, a B30.2/SPRY domain and a single RING-type zinc finger.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: RNF23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat