To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Family With Sequence Similarity 50, Member A
- Clonality:Polyclonal
- Gene ID:9130
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:FAM50A
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:HXC-26|XAP5|HXC26|9F|DXS9928E
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10477-950
- Supplier No.:BS-8152R
Specifications
About this item
FAM50A, also known as DXS9928E, HXC26, XAP5 or 9F, is a 339 amino acid nuclear protein that belongs to the FAM50 family. Expressed ubiquitously with highest expression in fetal kidney, liver and brain, as well as adult heart, spleen, skeletal muscle, prostate and small intestine, FAM50A is thought to function as a transcription factor that may bind to DNA. FAM50A contains an SV40 large T antigen nuclear localization signal and a polymorphic CCG repeat region in its 5’-UTR. Defects in the gene encoding FAM50A may be associated with acute lymphoblastic leukemia, suggesting a possible role for FAM50A in carcinogenesis.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: FAM50A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat