To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:2'-5'-Oligoadenylate Synthetase 3, 100kDa
- Clonality:Polyclonal
- Gene ID:4940
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:OAS3
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:p100|p100OAS
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10463-024
- Supplier No.:BS-7344R
Specifications
About this item
The 2'- 5'- oligoadenylate synthetase (OAS) family is comprised of four members: OAS1, OAS2, OAS3 and OASL. These proteins are induced by interferons and function to convert ATP into 2'- 5'- linked oligomers of adenosine in the presence of double-stranded RNA and magnesium ions. Copper, iron and zinc ions strongly inhibit the OAS enzymatic activity, while manganese ions can replace magnesium ions as an activator. The OAS family plays a significant role in the inhibition of cellular protein synthesis, apoptosis and growth, and its members are important factors in viral infection resistance. OAS3, also referred to as p100, contains three adjacent OAS1-like domains and maps to the human chromosome 12q24.2
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: OAS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat