To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 52 Member A3
- Clonality:Polyclonal
- Gene ID:113278
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:SLC52A3
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:hRFT2|C20orf54|BVVLS|RFVT3|bA371L19.1|BVVLS1|RFT2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at -20°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:446-469/469
- Purification:Purified by Protein A
- Cat. no.:10401-418
- Supplier No.:BS-4164R
Specifications
About this item
Transporter for riboflavin, which must be obtained as a nutrient via intestinal absorption. Riboflavin transport is Na(+)-independent at low pH but significantly reduced by Na(+) depletion under neutral pH conditions. Activity is strongly inhibited by riboflavin analogs, such as lumiflavin, flavin mononucleotide (FMN), flavin adenine dinucleotide (FAD), by methylene blue, and to a lesser extent by amiloride.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC52A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat