To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Erythroid Differentiation Regulatory Factor 1
- Clonality:Polyclonal
- Gene ID:26098
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:EDRF1
- Conjugation:Cy5®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:EDRF1_HUMAN.|Erythroid dferentiation related factor 1|Chromosome 10 open reading frame 137|EDRF1|C10orf137 protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10496-382
- Supplier No.:BS-9737R-CY5
Specifications
About this item
The tetratricopeptide repeat (TPR) motif is a degenerate, 34 amino acid sequence found in many proteins and acts to mediate protein-protein interactions in various pathways. At the sequence level, there can be up to 16 tandem TPR repeats, each of which has a helix-turn-helix shape that stacks on other TPR repeats to achieve ligand binding specificity. EDRF1 (erythroid differentiation-related factor 1), also known as C10orf137 (chromosome 10 open reading frame 137), is a 1,238 amino acid protein containing two TPR repeats. Localizing to nucleus, EDRF1 is involved in transcriptional activation of globin genes by regulating DNA-binding activity of GATA-1 transcription factor. EDRF1 may also play an important role in organ development and histological differentiation. EDRF1 exists as four alternatively spliced isoforms and is encoded by a gene mapping to human chromosome 10q26.13.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EDRF1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat