To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:RNA Binding Motif Protein 26
- Clonality:Polyclonal
- Gene ID:64062
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:RBM26
- Conjugation:Alexa Fluor® 647
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:ZC3H17|C13orf10|PPP1R132|SE70-2|PRO1777|ARRS2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10493-256
- Supplier No.:BS-9609R-A647
Specifications
About this item
Proteins containing RNA recognition motifs, including various hnRNP proteins, are implicated in the regulation of alternative splicing and protein components of snRNPs. The RBM (RNA-binding motif) gene family encodes proteins with an RNA binding motif that have been suggested to play a role in the modulation of apoptosis. RBM26 (RNA binding motif protein 26), whose alternative names include CTCL tumor antigen se70-2, C13orf10, ARRS2, SE70-2, ZC3H17, PRO1777, FLJ20957, RP11-255E21.1, MGC133295 or MGC133296, is a 1,007 amino acid protein with six isoforms which result due to alternative splicing. RBM26 also contains one C3H1-type zinc finger and two RRM (RNA recognition motif) domains. The gene encoding RBM26 maps to human chromosome 13q31.1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RBM26
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat