To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:hexamethylene bis-acetamide inducible 1
- Clonality:Polyclonal
- Gene ID:10614
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:HEXIM1
- Conjugation:Cy5®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:CLP1|MAQ1|HIS1|EDG1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10488-198
- Supplier No.:BS-9460R-CY5
Specifications
About this item
Hexamethylene bis-acetamide inducible 1 (HEXIM1) and Hexamethylene bis-acetamide inducible 2 (HEXIM2) comprise a family of proteins which inhibit positive transcription elongation factor b (P-TEFb) through association with 7SK. P-TEFb is composed of a catalytic subunit, Cdk9, and either Cyclin T1 or T2 as a regulatory subunit. This complex regulates eukaryotic gene expression at the level of elongation. The C-terminal domains of HEXIM proteins interact directly with each other. Via these domains, HEXIM1 and HEXIM2 form stable homo- and hetero-oligomers, which may aid in the formation of the 7SK small nuclear ribonucleic acid particle. Despite their similar functions, HEXIM1 and HEXIM2 exhibit distinct expression patterns in various established cell lines and human tissues.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HEXIM1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat