To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:5-Methylcytosine
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Multiple
- Antigen symbol:5-mC
- Conjugation:Cy5®
- ImmunoChemistry:Yes
- Size:100 µL
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Cat. no.:10490-056
- Supplier No.:BS-9450R-CY5
Specifications
About this item
Cytidine is a nucleoside formed by a cytosine attached to a ribose ring via a beta-N1-glycosidic bond. DNA is methylated on cytidines by DNA methylases (DNMTs)to generate 5-methylcytidine (5-mC), a potent epigenetics marker and regulator of gene expression. The reverse reaction (cytidine demethylation) starts with its oxidation to hydroxymethyl- (5-hmC), formyl- (5-fC), and carboxy- (5-caC) cytidine. Several enzymes, including the Tet family of proteins have been implicated in cytidine demethylation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: 5mC
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others